Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001321154.1 | 8416 | UTR 3 | NP_001308083.1 | ||
NM_001321155.1 | 8416 | UTR 3 | NP_001308084.1 | ||
NM_001321156.1 | 8416 | UTR 3 | NP_001308085.1 | ||
NM_001321157.1 | 8416 | UTR 3 | NP_001308086.1 | ||
NM_001321158.1 | 8416 | UTR 3 | NP_001308087.1 | ||
NM_001321159.1 | 8416 | UTR 3 | NP_001308088.1 | ||
NM_001321160.1 | 8416 | UTR 3 | NP_001308089.1 | ||
NM_001321161.1 | 8416 | UTR 3 | NP_001308090.1 | ||
NM_001321162.1 | 8416 | UTR 3 | NP_001308091.1 | ||
NM_024770.4 | 8416 | UTR 3 | NP_079046.2 | ||
XM_011511875.2 | 8416 | Intron | XP_011510177.1 | ||
XM_017004984.1 | 8416 | Intron | XP_016860473.1 | ||
XM_017004985.1 | 8416 | Intron | XP_016860474.1 | ||
XM_017004986.1 | 8416 | Intron | XP_016860475.1 | ||
XM_017004987.1 | 8416 | Intron | XP_016860476.1 | ||
XM_017004988.1 | 8416 | Intron | XP_016860477.1 | ||
XM_017004989.1 | 8416 | Intron | XP_016860478.1 |