Product Details

SNP ID
rs77768267
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:16787630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAAAGTTCACTAGACTCTATAA[C/G]GTATCCTGGGGAAGAACCACGCCAA
Phenotype
MIM: 612686
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
LOC283278 PubMed Links
Additional Information
For this assay, SNP(s) [rs74463290] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LOC283278
Gene Name
pleckstrin homology domain-containing family A member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017018657.1 Intron XP_016874146.1
XM_017018658.1 Intron XP_016874147.1
Gene
PLEKHA7
Gene Name
pleckstrin homology domain containing A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175058.4 Intron NP_778228.3
XM_017017240.1 Intron XP_016872729.1
XM_017017241.1 Intron XP_016872730.1
XM_017017242.1 Intron XP_016872731.1
XM_017017243.1 Intron XP_016872732.1

View Full Product Details