Product Details

SNP ID
rs376882956
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:154193706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTTGACATTTAGTTGGTTCCTA[A/G]TTAAACAACCCAATATCAAACATAT
Phenotype
MIM: 191030
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf189 PubMed Links

Gene Details

Gene
C1orf189
Gene Name
chromosome 1 open reading frame 189
There are no transcripts associated with this gene.

Gene
MIR190B
Gene Name
microRNA 190b
There are no transcripts associated with this gene.

Gene
TPM3
Gene Name
tropomyosin 3
There are no transcripts associated with this gene.

View Full Product Details