Product Details

SNP ID
rs553846338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18261911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCGGCAGCCGCCCCCTTCTGCGC[A/G]GTCATGCCGAGCCAGCACCTGGGCC
Phenotype
MIM: 104750
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAA1 PubMed Links

Gene Details

Gene
SAA1
Gene Name
serum amyloid A1
There are no transcripts associated with this gene.

Gene
ST13P5
Gene Name
suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) pseudogene 5
There are no transcripts associated with this gene.

View Full Product Details