Product Details

SNP ID
rs78203006
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1722954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGGGAGAGAGCAGACCAGAGCC[A/G]CTGTCCTGCTTTTCTGCCCTCTACT
Phenotype
MIM: 605303 MIM: 615975
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TACC3 PubMed Links

Gene Details

Gene
TACC3
Gene Name
transforming acidic coiled-coil containing protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006342.2 Intron NP_006333.1
XM_005247929.2 Intron XP_005247986.1
XM_005247930.2 Intron XP_005247987.1
XM_011513386.2 Intron XP_011511688.1
XM_017007653.1 Intron XP_016863142.1
Gene
TMEM129
Gene Name
transmembrane protein 129
There are no transcripts associated with this gene.

View Full Product Details