Product Details

SNP ID
rs7902934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96518310 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATAATTGTTTCCTTGGAACTCTG[C/T]ACCGACTGTCCATGCTCTGTGGGGA
Phenotype
MIM: 606743 MIM: 616872
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TLL2 PubMed Links

Gene Details

Gene
TLL2
Gene Name
tolloid like 2
There are no transcripts associated with this gene.

Gene
TM9SF3
Gene Name
transmembrane 9 superfamily member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020123.3 5940 UTR 3 NP_064508.3
XM_011539976.2 5940 Intron XP_011538278.1
XM_011539977.1 5940 UTR 3 XP_011538279.1

View Full Product Details