Product Details

SNP ID
hCV27848365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154295789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACATAAACGCACTGGGCAGCTC[A/G]CAGGCGCCATTCGCTCTTCAGACGC
Phenotype
MIM: 300092 MIM: 300044
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEX28 PubMed Links

Gene Details

Gene
TEX28
Gene Name
testis expressed 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205201.1 116 Intron NP_001192130.1
NM_001586.2 116 Intron NP_001577.1
Gene
TKTL1
Gene Name
transketolase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145933.1 116 UTR 5 NP_001139405.1
NM_001145934.1 116 Intron NP_001139406.1
NM_012253.3 116 UTR 5 NP_036385.3

View Full Product Details