Product Details

SNP ID
rs10420575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57633180 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCGCCCCCCCCAGGGCGGGACTT[C/G]TGGCGTCTTCGCAGCGGTCATTTTG
Phenotype
MIM: 604076 MIM: 601856
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF134 PubMed Links

Gene Details

Gene
ZNF134
Gene Name
zinc finger protein 134
There are no transcripts associated with this gene.

Gene
ZNF211
Gene Name
zinc finger protein 211
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001265597.2 14 UTR 5 NP_001252526.1
NM_001265598.2 14 UTR 5 NP_001252527.1
NM_001265599.2 14 UTR 5 NP_001252528.1
NM_001265600.2 14 UTR 5 NP_001252529.1
NM_001322306.1 14 UTR 5 NP_001309235.1
NM_006385.4 14 UTR 5 NP_006376.2
NM_198855.3 14 UTR 5 NP_942152.1

View Full Product Details