Product Details

SNP ID
rs45441892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43353721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCAGCCACAGACGGGTCATGAGC[C/G]CGGTATTACTGCTGGCCCTCCTGGG
Phenotype
MIM: 162860 MIM: 605157
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD177 PubMed Links

Gene Details

Gene
CD177
Gene Name
CD177 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020406.3 63 Missense Mutation CCG,GCG P3A NP_065139.2
XM_017027021.1 63 Missense Mutation CCG,GCG P3A XP_016882510.1
XM_017027022.1 63 Missense Mutation CCG,GCG P3A XP_016882511.1
Gene
PRG1
Gene Name
p53-responsive gene 1
There are no transcripts associated with this gene.

View Full Product Details