Product Details

SNP ID
rs555745012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27978884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTTCAAAGTCCTTTGCTCTCAG[C/G]GGGGGCTTTGATGTCTAAAGGGGCC
Phenotype
MIM: 600297 MIM: 615804
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDX2 PubMed Links

Gene Details

Gene
CDX2
Gene Name
caudal type homeobox 2
There are no transcripts associated with this gene.

Gene
URAD
Gene Name
ureidoimidazoline (2-oxo-4-hydroxy-4-carboxy-5-) decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105577.1 Intron NP_001099047.1

View Full Product Details