Product Details
- SNP ID
-
rs16827810
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
43
- Location
-
Chr.1:40981694 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGTTAAATGACTTGGTTTAATCAGC[A/G]TTTCTCTGATTTATATCCATAACAC
- Phenotype
-
MIM: 123860
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTPS1
PubMed Links
Gene Details
- Gene
- CTPS1
- Gene Name
- CTP synthase 1
View Full Product Details