Product Details
- SNP ID
-
rs145919939
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:86959206 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCAGGTCACGGATCCCCTCCCTCC[C/G]CAGAGAGAAGGGGCAGGCTGGGGGA
- Phenotype
-
MIM: 608925
MIM: 602998
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADIRF
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78801716] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADIRF
- Gene Name
- adipogenesis regulatory factor
There are no transcripts associated with this gene.
- Gene
- MMRN2
- Gene Name
- multimerin 2
- Gene
- SNCG
- Gene Name
- synuclein gamma
View Full Product Details