Product Details
- SNP ID
-
hCV27852471
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:45230771 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGCTGGGGCTATGGGGAGAGGG[C/G]CACCGCCATACTGCCCATGGAGCCA
- Phenotype
-
MIM: 604656
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FMNL1
PubMed Links
Gene Details
- Gene
- FMNL1
- Gene Name
- formin like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005892.3 |
|
Intron |
|
|
NP_005883.2 |
View Full Product Details