Product Details

SNP ID
rs3806714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186570776 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTAGCTAGCTGTCTCTGCGGACCA[A/G]GGAGACCCCCGCGCCCCCCCGGTGT
Phenotype
MIM: 611341
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB11 PubMed Links

Gene Details

Gene
DNAJB11
Gene Name
DnaJ heat shock protein family (Hsp40) member B11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016306.5 101 UTR 5 NP_057390.1
Gene
TBCCD1
Gene Name
TBCC domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134415.1 101 Intron NP_001127887.1
NM_001286749.1 101 Intron NP_001273678.1
NM_018138.4 101 Intron NP_060608.1

View Full Product Details