Product Details

SNP ID
hCV27853412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7354388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTATAGGTCCCTGGCATCCTGG[A/T]TCATATCTGTGTTGCTGACTACAGT
Phenotype
MIM: 607763 MIM: 609848
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ACAP1 PubMed Links

Gene Details

Gene
ACAP1
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014716.3 2500 Intron NP_055531.1
Gene
KCTD11
Gene Name
potassium channel tetramerization domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002914.2 2500 UTR 3 NP_001002914.1
Gene
TMEM95
Gene Name
transmembrane protein 95
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320435.1 2500 Intron NP_001307364.1
NM_001320436.1 2500 Intron NP_001307365.1
NM_198154.2 2500 Intron NP_937797.1

View Full Product Details