Product Details

SNP ID
rs116438665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103122841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCCGCTACTTAAAGTGGTGTGA[C/G]CCGGGTCAGCCACTTCACCTTTCCG
Phenotype
MIM: 603300
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00677 PubMed Links

Gene Details

Gene
LINC00677
Gene Name
long intergenic non-protein coding RNA 677
There are no transcripts associated with this gene.

Gene
TNFAIP2
Gene Name
TNF alpha induced protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006291.2 Intron NP_006282.2
XM_006720243.3 Intron XP_006720306.1
XM_011537112.2 Intron XP_011535414.1
XM_011537113.2 Intron XP_011535415.1
XM_011537114.2 Intron XP_011535416.1
XM_011537115.2 Intron XP_011535417.1
XM_017021616.1 Intron XP_016877105.1

View Full Product Details