Product Details
- SNP ID
-
hCV27856641
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:32887492 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTGCAGAACTGGCTGGCTGTTTC[C/T]TCTTCATTGCCTTCTACTCAGCCTC
- Phenotype
-
MIM: 142622
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HPCA
PubMed Links
Gene Details
- Gene
- HPCA
- Gene Name
- hippocalcin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002143.2 |
|
Intron |
|
|
NP_002134.2 |
- Gene
- TMEM54
- Gene Name
- transmembrane protein 54
There are no transcripts associated with this gene.
View Full Product Details