Product Details

SNP ID
rs17427882
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27186810 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCCAACAACTCTCTTCTTGGAAG[G/T]ATCTGGGATGTTGCTGAAGGAAAAT
Phenotype
MIM: 142957 MIM: 142958 MIM: 607530
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HOXA10 PubMed Links

Gene Details

Gene
HOXA10
Gene Name
homeobox A10
There are no transcripts associated with this gene.

Gene
HOXA10-HOXA9
Gene Name
HOXA10-HOXA9 readthrough
There are no transcripts associated with this gene.

Gene
HOXA11
Gene Name
homeobox A11
There are no transcripts associated with this gene.

Gene
HOXA11-AS
Gene Name
HOXA11 antisense RNA
There are no transcripts associated with this gene.

View Full Product Details