Product Details

SNP ID
hCV27861743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:37782169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCCACCTAAGTTCTTTTACACA[A/T]GAAAACTTCTTGGGGTAAGTATAAA
Phenotype
MIM: 300481
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYBB PubMed Links

Gene Details

Gene
CYBB
Gene Name
cytochrome b-245 beta chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000397.3 188 Nonsense Mutation AGA,TGA R43* NP_000388.2

View Full Product Details