Product Details

SNP ID
rs104894832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101398387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTTACCTGTCTAAGCTGGTACC[T/C]TTGCTTGCCCAAGGGGTCCTGATTG
Phenotype
MIM: 300300 MIM: 300644 MIM: 300610 MIM: 300902
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
BTK PubMed Links

Gene Details

Gene
BTK
Gene Name
Bruton tyrosine kinase
There are no transcripts associated with this gene.

Gene
GLA
Gene Name
galactosidase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000169.2 1092 Missense Mutation NP_000160.1
Gene
HNRNPH2
Gene Name
heterogeneous nuclear ribonucleoprotein H2 (H')
There are no transcripts associated with this gene.

Gene
RPL36A
Gene Name
ribosomal protein L36a
There are no transcripts associated with this gene.

Gene
RPL36A-HNRNPH2
Gene Name
RPL36A-HNRNPH2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199973.1 1092 Intron NP_001186902.1
NM_001199974.1 1092 Intron NP_001186903.1

View Full Product Details