Product Details
- SNP ID
-
rs4685054
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:13962816 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAATTCAAAGGATTTTAAAATTTTC[A/G]TCTTGATTTCAGTGTTAACCCAGAA
- Phenotype
-
MIM: 611167
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TPRXL
PubMed Links
Gene Details
- Gene
- TPRXL
- Gene Name
- tetrapeptide repeat homeobox like
There are no transcripts associated with this gene.
View Full Product Details