Product Details

SNP ID
hCV27876285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:3070976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAATCACTTAGCTCCAAATCTTA[G/T]TTTCTTTATTTGCAAATTGAAATCA
Phenotype
MIM: 603453
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RIPK1 PubMed Links

Gene Details

Gene
RIPK1
Gene Name
receptor interacting serine/threonine kinase 1
There are no transcripts associated with this gene.

View Full Product Details