Product Details
- SNP ID
-
rs4686476
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:188161812 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAATTATGGTATCTTAGGCTTGAAA[C/T]TTGCGCTTCATACTTAGAAAGCAAT
- Phenotype
-
MIM: 600700
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LPP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139439255] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LPP
- Gene Name
- LIM domain containing preferred translocation partner in lipoma
- Gene
- LPP-AS2
- Gene Name
- LPP antisense RNA 2
There are no transcripts associated with this gene.
View Full Product Details