Product Details

SNP ID
rs4996980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7690026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGTCCCCCAGCCCCCACTGGC[A/G]TCCTTCTCCCCTGCACCCTAGAGAC
Phenotype
MIM: 151445
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FCER2 PubMed Links

Gene Details

Gene
FCER2
Gene Name
Fc fragment of IgE receptor II
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207019.2 Intron NP_001193948.2
NM_001220500.1 Intron NP_001207429.1
NM_002002.4 Intron NP_001993.2
XM_005272462.4 Intron XP_005272519.1
Gene
TRAPPC5
Gene Name
trafficking protein particle complex 5
There are no transcripts associated with this gene.

View Full Product Details