Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018463.3 | 851 | Intron | NP_060933.3 | ||
XM_005253707.4 | 851 | Intron | XP_005253764.1 | ||
XM_005253709.4 | 851 | Intron | XP_005253766.1 | ||
XM_017019641.1 | 851 | Intron | XP_016875130.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_031474.2 | 851 | Intron | NP_113662.1 | ||
XM_005253797.3 | 851 | Intron | XP_005253854.1 | ||
XM_005253798.3 | 851 | Intron | XP_005253855.1 | ||
XM_011521031.2 | 851 | Intron | XP_011519333.1 | ||
XM_017020000.1 | 851 | Missense Mutation | ACT,AGT | T260S | XP_016875489.1 |
XM_017020001.1 | 851 | Intron | XP_016875490.1 |