Product Details

SNP ID
rs4517828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4148103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTTGCTGTAGGAGTTCAGGTGA[G/T]AAAGGATGGCAGCCTGGGCTTGCTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYB5D2 PubMed Links

Gene Details

Gene
CYB5D2
Gene Name
cytochrome b5 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001254755.1 Intron NP_001241684.1
NM_001254756.1 Intron NP_001241685.1
NM_144611.3 Intron NP_653212.1
Gene
ZZEF1
Gene Name
zinc finger ZZ-type and EF-hand domain containing 1
There are no transcripts associated with this gene.

View Full Product Details