Product Details

SNP ID
rs4533186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:59191152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAAGTGTGTATATGAGAGTGTC[C/T]GTGCATGTGTATCCATAGTCAGGTA
Phenotype
MIM: 606626
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DAAM1 PubMed Links

Gene Details

Gene
DAAM1
Gene Name
dishevelled associated activator of morphogenesis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270520.1 Intron NP_001257449.1
NM_014992.2 Intron NP_055807.1
XM_005267430.1 Intron XP_005267487.1
XM_005267431.1 Intron XP_005267488.1

View Full Product Details