Product Details
- SNP ID
-
rs4396712
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:227873396 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGATCATTGTAGCACCCTCAGCTT[A/C]TAGAAGAGATTTGGTGACACAGAAG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DAW1
PubMed Links
Gene Details
- Gene
- DAW1
- Gene Name
- dynein assembly factor with WD repeats 1
There are no transcripts associated with this gene.
View Full Product Details