Product Details

SNP ID
rs4434137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52313803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGAATGGAAGTATTTGGGGCAG[A/G]AAAAGTCTTGGTGTCCCTCACACCA
Phenotype
MIM: 603332
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAH1 PubMed Links

Gene Details

Gene
DNAH1
Gene Name
dynein axonemal heavy chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015512.4 Intron NP_056327.4

View Full Product Details