Product Details

SNP ID
rs4483957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49824033 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGACCGGTAAGACAGTCTTGAGTGA[C/T]GTTAAATGGAGACTTCTTGAGGCCC
Phenotype
MIM: 610622 MIM: 610197
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FUZ PubMed Links

Gene Details

Gene
FUZ
Gene Name
fuzzy planar cell polarity protein
There are no transcripts associated with this gene.

Gene
MED25
Gene Name
mediator complex subunit 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030973.3 Intron NP_112235.2
XM_011527353.1 Intron XP_011525655.1
Gene
MIR6800
Gene Name
microRNA 6800
There are no transcripts associated with this gene.

View Full Product Details