Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001317837.1 | 1006 | UTR 3 | NP_001304766.1 | ||
NM_001317838.1 | 1006 | UTR 3 | NP_001304767.1 | ||
NM_001317839.1 | 1006 | UTR 3 | NP_001304768.1 | ||
NM_001317841.1 | 1006 | UTR 3 | NP_001304770.1 | ||
NM_001317842.1 | 1006 | UTR 3 | NP_001304771.1 | ||
NM_152399.3 | 1006 | UTR 3 | NP_689612.3 | ||
XM_005262737.2 | 1006 | UTR 3 | XP_005262794.1 | ||
XM_017007725.1 | 1006 | UTR 3 | XP_016863214.1 | ||
XM_017007726.1 | 1006 | UTR 3 | XP_016863215.1 |