Product Details

SNP ID
rs4913078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140247095 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACTCAGGGAAGCAGGGAGAGTTT[A/T]GGGCAGAGGACAAGTGAGCTCTCTG
Phenotype
MIM: 604897
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYSTM1 PubMed Links

Gene Details

Gene
CYSTM1
Gene Name
cysteine rich transmembrane module containing 1
There are no transcripts associated with this gene.

Gene
PFDN1
Gene Name
prefoldin subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002622.4 Intron NP_002613.2
XM_005268465.3 Intron XP_005268522.1

View Full Product Details