Product Details

SNP ID
rs4447177
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:22624479 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTAATTCCATGGCTTTAAGTGGA[A/G]TACTTCCAATCACTTCCTCTATCCA
Phenotype
MIM: 613897 MIM: 602835
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FANCF PubMed Links
Additional Information
For this assay, SNP(s) [rs45473997] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FANCF
Gene Name
Fanconi anemia complementation group F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022725.3 1363 UTR 3 NP_073562.1
Gene
GAS2
Gene Name
growth arrest specific 2
There are no transcripts associated with this gene.

View Full Product Details