Product Details

SNP ID
rs4663772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:237631537 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATTCAGGCGGGATGTTATCAGGA[A/G]AAAGGTTTAGTACGCACTTGCAGAA
Phenotype
MIM: 603256
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRFIP1 PubMed Links

Gene Details

Gene
LRRFIP1
Gene Name
LRR binding FLII interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137550.1 Intron NP_001131022.1
NM_001137551.1 Intron NP_001131023.1
NM_001137552.1 Intron NP_001131024.1
NM_001137553.1 Intron NP_001131025.1
NM_004735.3 Intron NP_004726.2
XM_005246141.4 Intron XP_005246198.1
XM_005246142.2 Intron XP_005246199.1
XM_017005253.1 Intron XP_016860742.1
XM_017005254.1 Intron XP_016860743.1
XM_017005255.1 Intron XP_016860744.1
XM_017005256.1 Intron XP_016860745.1
XM_017005257.1 Intron XP_016860746.1
XM_017005258.1 Intron XP_016860747.1
XM_017005259.1 Intron XP_016860748.1
XM_017005260.1 Intron XP_016860749.1
XM_017005261.1 Intron XP_016860750.1
XM_017005262.1 Intron XP_016860751.1
XM_017005263.1 Intron XP_016860752.1

View Full Product Details