Product Details

SNP ID
rs4262008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:75737153 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTCAAACTACTTGGCTGAATGCT[A/G]TCTCACTACTTACTTTATAAATGTG
Phenotype
MIM: 603344
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
USO1 PubMed Links

Gene Details

Gene
USO1
Gene Name
USO1 vesicle transport factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290049.1 Intron NP_001276978.1
NM_003715.3 Intron NP_003706.2
XM_006714396.3 Intron XP_006714459.1
XM_006714397.3 Intron XP_006714460.1

View Full Product Details