Product Details

SNP ID
rs4804194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12461486 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTGCACAGTGCAGTGGAGAAGCA[A/G]ACACCCATGTAGTTCTTTTTGACTG
Phenotype
MIM: 606697
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF443 PubMed Links

Gene Details

Gene
ZNF443
Gene Name
zinc finger protein 443
There are no transcripts associated with this gene.

Gene
ZNF709
Gene Name
zinc finger protein 709
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152601.3 4597 UTR 3 NP_689814.1

View Full Product Details