Product Details

SNP ID
rs4949356
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:31181141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAATTGAGTTAAATGAGAAGTCT[A/G]AAAGAGTCCATTTGCTAAGGGATTA
Phenotype
MIM: 612871
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NKAIN1 PubMed Links

Gene Details

Gene
NKAIN1
Gene Name
Na+/K+ transporting ATPase interacting 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024522.2 1526 UTR 3 NP_078798.2
XM_017002320.1 1526 UTR 3 XP_016857809.1

View Full Product Details