Product Details

SNP ID
rs4404900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:58563262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTATTCTGCTATGAAAATGAAT[A/G]AAATACTGATACATGCTCCAACACA
Phenotype
MIM: 602217
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDCBP PubMed Links

Gene Details

Gene
SDCBP
Gene Name
syndecan binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007067.1 Intron NP_001007068.1
NM_001007068.1 Intron NP_001007069.1
NM_001007069.1 Intron NP_001007070.1
NM_001007070.1 Intron NP_001007071.1
NM_005625.3 Intron NP_005616.2

View Full Product Details