Product Details

SNP ID
hCV27971331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21389140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCCAGGTGTTAGGCTGTTTCA[G/A]AATTCCAGAGGACGGGGTCTGAGTC
Phenotype
MIM: 602323
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
KCNJ12 PubMed Links

Gene Details

Gene
KCNJ12
Gene Name
potassium voltage-gated channel subfamily J member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021012.4 Intron NP_066292.2

View Full Product Details