Product Details
- SNP ID
-
rs4824800
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:49687706 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTAAGGGCGATCCTGTGTCTTATA[A/G]CCAAGTAAACCTCTGTATAAATTTT
- Phenotype
-
MIM: 300288
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PAGE1
PubMed Links
Gene Details
- Gene
- PAGE1
- Gene Name
- PAGE family member 1
View Full Product Details