Product Details

SNP ID
rs4770676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:24429745 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGATTCCTTGAGGTTGCACCTGT[A/G]TATGTAGAGGTTAGGAGACAGCCAA
Phenotype
MIM: 607519
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PARP4 PubMed Links

Gene Details

Gene
PARP4
Gene Name
poly(ADP-ribose) polymerase family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006437.3 Intron NP_006428.2
XM_011534931.1 Intron XP_011533233.1
XM_011534932.2 Intron XP_011533234.1

View Full Product Details