Product Details

SNP ID
rs4605278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51519690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCCTTCTAAAATTCATATGTTG[A/G]AAGTCCTAAATCTCAATATCTCAGA
Phenotype
MIM: 604405
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIGLEC6 PubMed Links

Gene Details

Gene
SIGLEC6
Gene Name
sialic acid binding Ig like lectin 6
There are no transcripts associated with this gene.

View Full Product Details