Product Details

SNP ID
rs4454968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55509018 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCCAGAGCCTGGGGCGTGTCACA[G/T]CCGAAGGGGCAGGGCGGCAGCAGCA
Phenotype
MIM: 612040 MIM: 120360
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LPCAT2 PubMed Links

Gene Details

Gene
LPCAT2
Gene Name
lysophosphatidylcholine acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017839.4 18 UTR 5 NP_060309.2
Gene
MMP2
Gene Name
matrix metallopeptidase 2
There are no transcripts associated with this gene.

View Full Product Details