Product Details

SNP ID
rs4145896
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:160543023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGTGGAAATTTACAGTTTCTCT[C/T]ATTTTATATTAACTCATCAGAATGT
Phenotype
MIM: 604513
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD84 PubMed Links

Gene Details

Gene
CD84
Gene Name
CD84 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184879.1 5989 UTR 3 NP_001171808.1
NM_001184881.1 5989 UTR 3 NP_001171810.1
NM_001184882.1 5989 UTR 3 NP_001171811.1
NM_003874.3 5989 UTR 3 NP_003865.1
XM_011510094.2 5989 UTR 3 XP_011508396.1
XM_011510095.2 5989 UTR 3 XP_011508397.1

View Full Product Details