Product Details

SNP ID
hCV27998111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43194713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACACTGTTTCAAGTAGGTACTA[C/G]AACTGATCACCCGTGAAGTGCACGT
Phenotype
MIM: 616616 MIM: 614531
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CSGALNACT2 PubMed Links

Gene Details

Gene
CSGALNACT2
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 2
There are no transcripts associated with this gene.

Gene
RASGEF1A
Gene Name
RasGEF domain family member 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282862.1 3058 UTR 3 NP_001269791.1
NM_145313.3 3058 UTR 3 NP_660356.2

View Full Product Details