Product Details

SNP ID
rs4932145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89833227 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCAGTTAAATGGAGGGACACAGAC[A/G]ACTCTTTGGCTCTTATACTCTATGA
Phenotype
MIM: 602416
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP3S2 PubMed Links

Gene Details

Gene
AP3S2
Gene Name
adaptor related protein complex 3 sigma 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005829.4 3306 UTR 3 NP_005820.1
Gene
C15orf38-AP3S2
Gene Name
C15orf38-AP3S2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199058.1 3306 UTR 3 NP_001185987.1

View Full Product Details