Product Details

SNP ID
rs4674238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218036816 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTCTGGTTTGCAGTTTAAATATC[G/T]CTGGTTATGGCATCAGGTATTTTGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RUFY4 PubMed Links

Gene Details

Gene
RUFY4
Gene Name
RUN and FYVE domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198483.3 Intron NP_940885.2
XM_011511014.2 Intron XP_011509316.1
XM_017003895.1 Intron XP_016859384.1
XM_017003896.1 Intron XP_016859385.1
XM_017003897.1 Intron XP_016859386.1

View Full Product Details