Product Details

SNP ID
rs4542469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14833821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGGCAAATGCTATCTATAGCCT[C/G]TTTCTGTCAAAATGCAAGGAGCAAG
Phenotype
MIM: 110600
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ART4 PubMed Links

Gene Details

Gene
ART4
Gene Name
ADP-ribosyltransferase 4 (Dombrock blood group)
There are no transcripts associated with this gene.

Gene
C12orf60
Gene Name
chromosome 12 open reading frame 60
There are no transcripts associated with this gene.

View Full Product Details