Product Details

SNP ID
rs4850874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:98453336 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCACACAGCATGTTTCAAAATGCA[A/G]TGGTTCATGAAATCAATTTGAGGTT
Phenotype
MIM: 600916
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INPP4A PubMed Links

Gene Details

Gene
INPP4A
Gene Name
inositol polyphosphate-4-phosphatase type I A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134224.1 Intron NP_001127696.1
NM_001134225.1 Intron NP_001127697.1
NM_001566.2 Intron NP_001557.1
NM_004027.2 Intron NP_004018.1
XM_006712499.3 Intron XP_006712562.1
XM_006712500.3 Intron XP_006712563.1
XM_006712501.3 Intron XP_006712564.1
XM_006712502.3 Intron XP_006712565.1
XM_006712503.3 Intron XP_006712566.1
XM_006712504.3 Intron XP_006712567.1
XM_006712505.3 Intron XP_006712568.1
XM_006712506.3 Intron XP_006712569.1
XM_006712507.3 Intron XP_006712570.1
XM_006712508.3 Intron XP_006712571.1
XM_011511122.2 Intron XP_011509424.1
XM_011511123.2 Intron XP_011509425.1
XM_011511124.2 Intron XP_011509426.1
XM_011511125.2 Intron XP_011509427.1
XM_011511127.2 Intron XP_011509429.1
XM_017003993.1 Intron XP_016859482.1
XM_017003994.1 Intron XP_016859483.1
XM_017003995.1 Intron XP_016859484.1
XM_017003996.1 Intron XP_016859485.1
XM_017003997.1 Intron XP_016859486.1
XM_017003998.1 Intron XP_016859487.1
XM_017003999.1 Intron XP_016859488.1
XM_017004000.1 Intron XP_016859489.1
XM_017004001.1 Intron XP_016859490.1
XM_017004002.1 Intron XP_016859491.1
XM_017004003.1 Intron XP_016859492.1

View Full Product Details