Product Details

SNP ID
rs7058353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:72181764 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTACGGCAACTGGAGCGGTTCAG[A/C]GTTCAACAACAAGCTTCCAAGATGC
Phenotype
MIM: 300252
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PIN4 PubMed Links

Gene Details

Gene
PIN4
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170747.1 89 Missense Mutation AGA,AGC R18S NP_001164218.1
NM_006223.3 89 Missense Mutation AGA,AGC R18S NP_006214.2

View Full Product Details